Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AMIGO3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544452
Description
Peptide sequence: MTWLVLLGTL LCMLRVGLGT PDSEGFPPRA LHNCPYKCIC AADLLSCTGL Sequence homology: Guinea Pig: 79%; Human: 100%.
Amphoterin induced gene and ORF 3 (AMIGO3) is a gene located on chromosome 3p21.31, encoding a member of the AMIGO family of proteins involved in cell adhesion, axon guidance, and neuronal signaling. AMIGO3 is primarily expressed in the central nervous system, playing a crucial role in neural development and the maintenance of neural circuits. Functionally, AMIGO3 contributes to neuronal growth and synapse formation, impacting processes such as neural plasticity and signal transduction. The protein's activity facilitates interactions with other neuronal surface proteins, influencing axon pathfinding and connectivity. Aberrant expression or mutations in AMIGO3 have been linked to neurological disorders and neurodegenerative diseases, suggesting its significance in neural health and pathology. Research into AMIGO3 continues to explore its potential therapeutic applications in promoting neural repair and modulating synaptic functions, offering insights into strategies for addressing nervous system disorders.
Specifications
AMIGO3 | |
Polyclonal | |
Unconjugated | |
Amigo3 | |
adhesion molecule with Ig like domain 3; adhesion molecule with Ig-like domain 3; ALI3; alivin 3; alivin-3; AMIGO3; AMIGO-3; amigo3 {ECO:0000312; amphoterin induced gene and ORF 3; amphoterin-induced gene and ORF 3; amphoterin-induced protein 3; E430002N15Rik; EMBL:AAO48952.1, ECO:0000312; Kiaa1851; MGC120552; mKIAA1851; RGD:631413}; transmembrane protein AMIGO3; UNQ6084/PRO20089 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
386724 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86WK7 | |
Amigo3 | |
Synthetic peptide directed towards the N-terminal of human AMIGO3 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction