Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMIGO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | AMIGO3 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3347
![]() ![]() |
Proteintech Group Inc
212781AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
AMIGO3 may mediate heterophilic cell-cell interaction. AMIGO3 may contribute to signal transduction through its intracellular domain.Specifications
AMIGO3 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q80ZD5, Q86WK7, Q8C2S7 | |
316003, 320844, 386724 | |
AMIGO3 Fusion Protein Ag15790 | |
Primary | |
-20°C | |
AMIGO3 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ALI3, Alivin 3, AMIGO 3, AMIGO3, Amphoterin induced protein 3, KIAA1851 | |
Amigo3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title