Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amino-2-Naphthol-4-Sulfonic Acid, 0.25% w/v APHA 4500, ASTM D859-05, Reagents
Amino-2-Naphthol-4-Sulfonic Acid, 0.25% w/v APHA 4500, ASTM D859-05, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CA215000500B
Quantity | 500 mL |
---|
Chemical Identifiers
7732-18-5, 7631-90-5, 7631-90-5 | |
18.02 | |
ANSA | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
Colorless | |
100°C | |
ANSA | |
O | |
18.02 | |
1.05 | |
Non-sterile | |
Amino-2-Naphthol-4-Sulfonic Acid, 0.25% w/v APHA 4500, ASTM D859-05 |
7732-18-5, 7631-90-5, 7631-90-5 | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
500 mL | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction