Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Amino Acid Standard H PROMO

Catalog No. PI20088PM
Change view
Click to view available options
:
Thermo Scientific™ Amino Acid Standard H
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PI20088PM Thermo Scientific™ Amino Acid Standard H
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20088PM Supplier Thermo Scientific™ Supplier No. 20088

Calibrate HPLC analysis of protein hydrolysates using this calibration standard mixture of 18 amino acids, each at 2.5 micromoles/mL in 0.1N HCl.

Highlights:

  • Uses L-form configuration to permit standardization of microbial and other assays
  • Molar concentration verified by conventional amino acid analysis methods
  • With the exception of cystine, each amino acid is supplied at a concentration of 2.5μmoles/mL in 0.1N HCl
  • L-alanine, Ammonia [(NH4)2SO4], L-Arginine, L-Aspartic Acid, L-Cystine, L-Glutamic Acid, Glycine, L-Histidine, L-Isoleucine, L-Leucine, L-Lysine·HCl, L-Methionine, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tyrosine, L-Valine

Includes:

  • L-alanine, Ammonia [(NH4)2SO4], L-Arginine, L-Aspartic Acid, L-Cystine, L-Glutamic Acid, Glycine, L-Histidine, L-Isoleucine, L-Leucine, L-Lysine·HCl, L-Methionine, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tyrosine, L-Valine

Specifications

Product Type Amino Acid Standard H
Content And Storage Store between -20°C and 8°C.
Quantity 10 x 1 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.