Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amino Acid Standards, Physiological, Analytical Standard, Acidics and Neutrals, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL
Specifications
Specifications
Packaging | Glass Bottle |
Physical Form | Neat |
MDL Number | MFCD00162052 |
Quantity | 5 mL |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction