Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminoacylase/ACY1 Antibody (128), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289559PE
Description
Aminoacylase/ACY1 Monoclonal antibody specifically detects Aminoacylase/ACY1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Aminoacylase/ACY1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 128 | |
| Immunohistochemistry-Paraffin | |
| ACY-1, ACY1D, acylase, aminoacylase 1, aminoacylase-1, EC 3.5.1, EC 3.5.1.14, N-acyl-L-amino-acid amidohydrolase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Aminoacylase/ACY1 (Accession#: NP_000657.1; Met1-Ser408). | |
| 0.1 mL | |
| metabolism, Proteases & Other Enzymes | |
| 95 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction