Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminoacylase/ACY1 Antibody (OTI1D4), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270080AF647
Description
Aminoacylase/ACY1 Monoclonal specifically detects Aminoacylase/ACY1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Aminoacylase/ACY1 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ACY1 | |
Full length recombinant protein of human Aminoacylase/ACY1 (NP_000657) produced in HEK293 cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1D4 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
ACY-1, ACY1D, acylase, aminoacylase 1, aminoacylase-1, EC 3.5.1, EC 3.5.1.14, N-acyl-L-amino-acid amidohydrolase | |
Mouse | |
Affinity Purified | |
metabolism, Proteases & Other Enzymes | |
95.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction