Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aminoacylase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583314
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583314 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583314 Supplier Invitrogen™ Supplier No. PA583314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGMELFVQRP EFHALRAGFA LDEGIANPTD AFTVFYSERS PWWVRVTSTG RPGHASRFME DTAAEKLHKV VNSILAFRE Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 91%.

ACY1 encodes a cytosolic, homodimeric, zinc-binding enzyme that catalyzes the hydrolysis of acylated L-amino acids to L-amino acids and an acyl group, and has been postulated to function in the catabolism and salvage of acylated amino acids. This gene is located on chromosome 3p21. 1, a region reduced to homozygosity in small-cell lung cancer (SCLC), and its expression has been reported to be reduced or undetectable in SCLC cell lines and tumors. The amino acid sequence of human aminoacylase-1 is highly homologous to the porcine counterpart, and this enzyme is the first member of a new family of zinc-binding enzymes. Mutations in this gene cause aminoacylase-1 deficiency, a metabolic disorder characterized by central nervous system defects and increased urinary excretion of N-acetylated amino acids. Alternative splicing of this gene results in multiple transcript variants. Read-through transcription also exists between this gene and the upstream ABHD14A (abhydrolase domain containing 14A) gene. A related pseudogene has been identified on chromosome 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aminoacylase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACY1
Gene Accession No. Q03154
Gene Alias 1110014J22Rik; ACY IA; ACY IB; ACY1; ACY-1; Acy1a; ACY-1A; Acy1b; ACY-1B; ACY1D; acylase; aminoacylase 1; aminoacylase I; aminoacylase-1; Aminoacylase-1A; aminoacylase-1B; epididymis secretory protein Li 5; HEL-S-5; N-acylamino acid aminohydrolase (Aminoacylase 1); N-acyl-L-amino-acid amidohydrolase
Gene Symbols ACY1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACY1. Recombinant protein control fragment (Product #RP-96318).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 95
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.