Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aminoacylase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589244
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589244 Supplier Invitrogen™ Supplier No. PA589244
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTSKGPEEEH PSVTLFRQYL RIRTVQPKPD YGAAVAFFEE TARQLGLGCQ KVEVAPGYVV TVLTWPGTNP TLSSILLNSH TDVVPVFKEH WSHDPFEAFK DSEGYIYARG AQDMKCVSIQ YLEAVRRLKV EGHRFPRTIH MTFVPDEEVG GHQGMELFVQ RPEFHALRAG FALDEGIANP TDAFTVFYSE RSPWWVRVTS TGRPGHASRF MEDTAAEKLH KVVNSILAFR EKEWQRLQSN PHLKEGSVTS VNLTKLEGGV AYNVIPATMS ASFDFRVAPD VDFKAFEEQL QSWCQAAGEG VTLEFAQKWM HPQVTPTDDS NPWWAAFSRV CKDMNLTLEP EIMPAATDNR YIRAVGVPAL GFSPMNRTPV LLHDHDERLH EAVFLRGVDI YTRLLPALAS VPALPSDS; Positive Samples: HepG2, U-251MG, A-431, Mouse kidney, Rat kidney; Cellular Location: Cytoplasm.

ACY1 encodes a cytosolic, homodimeric, zinc-binding enzyme that catalyzes the hydrolysis of acylated L-amino acids to L-amino acids and an acyl group, and has been postulated to function in the catabolism and salvage of acylated amino acids. This gene is located on chromosome 3p21. 1, a region reduced to homozygosity in small-cell lung cancer (SCLC), and its expression has been reported to be reduced or undetectable in SCLC cell lines and tumors. The amino acid sequence of human aminoacylase-1 is highly homologous to the porcine counterpart, and this enzyme is the first member of a new family of zinc-binding enzymes. Mutations in this gene cause aminoacylase-1 deficiency, a metabolic disorder characterized by central nervous system defects and increased urinary excretion of N-acetylated amino acids. Alternative splicing of this gene results in multiple transcript variants. Read-through transcription also exists between this gene and the upstream ABHD14A (abhydrolase domain containing 14A) gene. A related pseudogene has been identified on chromosome 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aminoacylase
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACY1
Gene Accession No. Q03154, Q6AYS7, Q99JW2
Gene Alias 1110014J22Rik; ACY IA; ACY IB; ACY1; ACY-1; Acy1a; ACY-1A; Acy1b; ACY-1B; ACY1D; acylase; aminoacylase 1; aminoacylase I; aminoacylase-1; Aminoacylase-1A; aminoacylase-1B; epididymis secretory protein Li 5; HEL-S-5; N-acylamino acid aminohydrolase (Aminoacylase 1); N-acyl-L-amino-acid amidohydrolase
Gene Symbols ACY1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-408 of human ACY1 (NP_0011858241).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109652, 300981, 95
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.