Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoguanidine bicarbonate, 98.5%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic Bottle
Plastic drum
2582-30-1
CH6N4·H2CO3
136.11
OTXHZHQQWQTQMW-UHFFFAOYSA-N
aminoguanidine bicarbonate,aminoguanidine hydrogen carbonate,aminoguanidinium bicarbonate,aminoguanidine hydrocarbonate,aminoguanidine carbonate 1:1,aminoguanidium hydrogen carbonate,n1-aminoguanidine carbonate 1:1,aminoguanidine hydrogencarbonate,aminoguanidinebicarbonate,hydrazinecarboximidamide carbonate
164944
2-aminoguanidine;carbonic acid
C(=NN)(N)N.C(=O)(O)O
2582-30-1
CH6N4·H2CO3
136.11
OTXHZHQQWQTQMW-UHFFFAOYSA-N
aminoguanidine bicarbonate,aminoguanidine hydrogen carbonate,aminoguanidinium bicarbonate,aminoguanidine hydrocarbonate,aminoguanidine carbonate 1:1,aminoguanidium hydrogen carbonate,n1-aminoguanidine carbonate 1:1,aminoguanidine hydrogencarbonate,aminoguanidinebicarbonate,hydrazinecarboximidamide carbonate
164944
2-aminoguanidine;carbonic acid
C(=NN)(N)N.C(=O)(O)O
Infrared Spectrum Authentic
Melting Point 125°C
Residue after Ignition 0.3% max
Color White
Quantity 250g
Packaging Plastic Bottle
Ignition Residue 0.3% max.
Assay Percent Range 98.5%
Linear Formula H2NNHC(=NH)NH2·H2CO3
Beilstein 03,117
Solubility Information Solubility in water: <5g/L (20°C)
Formula Weight 136.11
Physical Form Crystalline Powder
Percent Purity 98.50%
Chemical Name or Material Aminoguanidine bicarbonate
Show More Show Less

GHS H Statement
Toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May damage fertility or the unborn child.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention

GHS Signal Word: Danger

EINECSNumber : 219-956-7