Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoguanidine hydrogen carbonate, 98+%

Catalog No. p-7027718
Click to view available options
Quantity:
500 g
2500 g

Catalog No. AAB2469636

Supplier: Thermo Scientific Chemicals B2469636

Only null left
Add to Cart

CAS: 2582-30-1 | C2H8N4O3 | 136.111 g/mol

Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.

Solubility
Soluble in water (3.3 mg/ml at 30°C), and water (2.7 mg/ml at 20°C).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2582-30-1
Molecular Formula C2H8N4O3
Molecular Weight (g/mol) 136.111
MDL Number MFCD00012949
InChI Key OTXHZHQQWQTQMW-UHFFFAOYSA-N
Synonym aminoguanidine bicarbonate, aminoguanidine hydrogen carbonate, aminoguanidinium bicarbonate, aminoguanidine hydrocarbonate, aminoguanidine carbonate 1:1, aminoguanidium hydrogen carbonate, n1-aminoguanidine carbonate 1:1, aminoguanidine hydrogencarbonate, aminoguanidinebicarbonate, hydrazinecarboximidamide carbonate
PubChem CID 164944
IUPAC Name 2-aminoguanidine;carbonic acid
SMILES C(=NN)(N)N.C(=O)(O)O

Specifications

Melting Point ∼170°C (decomposition)
Density 1.6
Quantity 500 g
UN Number UN3077
Beilstein 3569869
Solubility Information Soluble in water (3.3mg/ml at 30°C),and water (2.7mg/ml at 20°C).
Formula Weight 136.11
Percent Purity ≥98%
Chemical Name or Material Aminoguanidine hydrogen carbonate
Hazard Category H317-H360
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 219-956-7
RTECSNumber FG1772000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.