Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

δ-Aminolevulinic Acid, 98%, MP Biomedicals

Catalog No. icn19462125
Click to view available options
Quantity:
25 mg

Chemical Identifiers

CAS 7222-1-29
Molecular Formula C5H10ClNO3
Molecular Weight (g/mol) 167.59
InChI Key ZLHFONARZHCSET-UHFFFAOYSA-N

Specifications

Quantity 25 mg
Synonym 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
SMILES C(CC(=O)O)C(=O)CN.Cl
IUPAC Name 5-amino-4-oxopentanoic acid;hydrochloride
PubChem CID 123608
Formula Weight 167.6
Grade Cell Culture Reagent
Chemical Name or Material δ-Aminolevulinic Acid, 98%, Hydrochloride
Recommended Storage 0°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.