Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminomethanesulfonic Acid 97.0+%, TCI America™
SDP

Catalog No. A0852250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0852250G 250 g
A085225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0852250G Supplier TCI America Supplier No. A0852250G

Chemical Identifiers

CAS 13881-91-9
Molecular Formula CH5NO3S
Molecular Weight (g/mol) 111.115
MDL Number MFCD00008124
InChI Key OBESRABRARNZJB-UHFFFAOYSA-N
Synonym methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid
PubChem CID 83791
IUPAC Name aminomethanesulfonic acid
SMILES C(N)S(=O)(=O)O

Specifications

Color White
Quantity 250 g
UN Number 3261
Formula Weight 111.12
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aminomethanesulfonic Acid
EINECSNumber (2)-3405
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.