Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminomethyltransferase Antibody (OTI6F4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270171PCP
Description
Aminomethyltransferase Monoclonal antibody specifically detects Aminomethyltransferase in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Aminomethyltransferase | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
OTI6F4 | |
Western Blot, Immunohistochemistry | |
aminomethyltransferase, EC 2.1.2.10, GCE, GCSTaminomethyltransferase (glycine cleavage system protein T), glycine cleavage system protein T, NKHmitochondrial | |
Human recombinant protein fragment corresponding to amino acids 146-403 of human AMT(NP_000472) produced in E.coli. | |
0.1 mL | |
Neuroscience | |
275 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction