Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminopeptidase A/ENPEP Antibody (6C3), Alexa Fluor™ 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143281AF594
Description
Aminopeptidase A/ENPEP Monoclonal specifically detects Aminopeptidase A/ENPEP in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Aminopeptidase A/ENPEP | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
ENPEP | |
The immunogen for this antibody was Aminopeptidase A/APA. | |
0.1 mL | |
Primary | |
Mouse | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
6C3 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Aminopeptidase A, APA, AP-A, CD249, CD249 antigen, Differentiation antigen gp160, EAP, EC 3.4.11, EC 3.4.11.7, glutamyl aminopeptidase, glutamyl aminopeptidase (aminopeptidase A), gp160 | |
Rat | |
Protein A or G purified | |
Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
2028.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction