Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminopeptidase A/ENPEP Antibody (SR1222), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Aminopeptidase A/ENPEP |
---|---|
Clone | SR1222 |
Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
Aminopeptidase A/ENPEP Monoclonal antibody specifically detects Aminopeptidase A/ENPEP in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Aminopeptidase A/ENPEP | |
Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Aminopeptidase A, APA, AP-A, CD249, CD249 antigen, Differentiation antigen gp160, EAP, EC 3.4.11, EC 3.4.11.7, glutamyl aminopeptidase, glutamyl aminopeptidase (aminopeptidase A), gp160 | |
A synthesized peptide derived from human Aminopeptidase A/ENPEP (Uniprot #: Q07075) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1222 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2028 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title