Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminopeptidase B/RNPEP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Aminopeptidase B/RNPEP |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Aminopeptidase B/RNPEP Polyclonal specifically detects Aminopeptidase B/RNPEP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Aminopeptidase B/RNPEP | |
Polyclonal | |
Rabbit | |
Human | |
aminopeptidase B, APB, AP-B, Arginine aminopeptidase, Arginyl aminopeptidase, arginyl aminopeptidase (aminopeptidase B), DKFZp547H084, EC 3.4.11, EC 3.4.11.6 | |
RNPEP | |
IgG | |
Affinity Purified |
Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
6051 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KNDHQEDFWKVKEFLHNQGKQKYTLPLYHAMMGGSEVAQTLAKETFASTASQLHSNVVNYVQQIVAPK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title