Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminopeptidase B/RNPEP Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259130
Description
Aminopeptidase B/RNPEP Polyclonal specifically detects Aminopeptidase B/RNPEP in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Aminopeptidase B/RNPEP | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
aminopeptidase B, APB, AP-B, Arginine aminopeptidase, Arginyl aminopeptidase, arginyl aminopeptidase (aminopeptidase B), DKFZp547H084, EC 3.4.11, EC 3.4.11.6 | |
Rabbit | |
Affinity Purified | |
RUO | |
6051 | |
Store at 4C. |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
RNPEP | |
Maps to a region between 600 and 650 | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction