Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminopeptidase LRAP/ERAP2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281783
Description
Aminopeptidase LRAP/ERAP2 Polyclonal specifically detects Aminopeptidase LRAP/ERAP2 in Human samples. It is validated for Western Blot, ELISA.Specifications
Aminopeptidase LRAP/ERAP2 | |
Polyclonal | |
Unconjugated | |
PBS | |
ERAP2 | |
ERAP2 antibody was raised against a 17 amino acid peptide near the amino terminus of human ERAP2. The immunogen is located within amino acids 110 - 160 of ERAP2 . Amino Acid Squence: LQSEEDSRYMKPGKELK | |
Affinity Purified | |
RUO | |
64167 | |
Human | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1-2 ug/mL, ELISA | |
EC 3.4.11, EC 3.4.11.3, endoplasmic reticulum aminopeptidase 2, FLJ23633, FLJ23701, FLJ23807, Leukocyte-derived arginine aminopeptidase, LRAPL-RAPEC 3.4.11.- | |
Rabbit | |
120 kDa | |
0.1 mg | |
Primary | |
Aminopeptidase LRAP/ERAP2 antibody is human reactive. Multiple isoforms of Aminopeptidase LRAP/ERAP2 are known to exist. Aminopeptidase LRAP/ERAP2 antibody is predicted to not cross-react with ERAP1. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction