Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminopeptidase N (CD13), Mouse anti-Human, Clone: 452, Millipore Sigma™

Catalog No. MABF2147100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2147100 100 μg
MABF214725 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2147100 Supplier MilliporeSigma Supplier No. MABF2147100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Aminopeptidase N (CD13), clone 452, Cat. No. MABF2147, is a mouse monoclonal antibody that detects Aminopeptidase N and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Agonist function, and Western Blotting.

Aminopeptidase N (UniProt: P15144; also known as EC:3.4.11.2, AP-N, hAPN, Alanyl aminopeptidase, Aminopeptidase M, AP-M, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, gp150, CD13) is encoded by the ANPEP (also known as APN, CD13, PEPN) gene (Gene ID: 290) in human. Aminopeptidase N is a single-pass type II membrane protein that is also found as a soluble form in cells. It is widely expressed as a homodimer of 280 kDa on the cell surface in many tissues, including intestinal epithelia and the nervous system. It serves as a broad specificity aminopeptidase that plays a role in the final digestion of peptides generated from hydrolysis of proteins by gastric and pancreatic proteases. It preferentially cleaves N-terminus neutral amino acids, most notably alanine residue. Aminopeptidase N is involved in many physiological processes, including antigen presentation regulation, differentiation, proliferation, apoptosis, cancer metastasis, and angiogenesis. It is also involved in the processing of various peptides including peptide hormones, such as angiotensin III and IV, neuropeptides, and chemokines. Human Aminopeptidase N consists of a short N-terminal cytoplasmic end (aa 2-8), a transmembrane domain (aa 9-32), and a large extracellular portion (aa 33-967), which is composed of a Ser/Thr-rich region (aa 33-68) and the metalloprotease domain (aa 69-967).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aminopeptidase N (CD13)
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibition Assays, Western Blot
Classification Monoclonal
Clone 452
Conjugate Unconjugated
Dilution Immunocytochemistry Analysis: A representative lot detected Aminopeptidase N (CD13) in Immunocytochemistry applications (Rahman, M.M., et. al. (2014). Front Physiol. 4:402; Subramani, J., et. al. (2013). J Immunol. 191(7):3905-12).Flow Cytometry Analysis: A representative lot detected Aminopeptidase N (CD13) in Flow Cytometry applications (Subramani, J., et. al. (2013). J Immunol. 191(7):3905-12).Western Blotting Analysis: A representative lot detected Aminopeptidase N (CD13) in Western Blotting applications (Rahman, M.M., et. al. (2014). Stem Cells. 32(6):1564-77; Rahman, M.M., et. al. (2014). Front Physiol. 4:402).Agonist Analysis: A representative lot induced a temporal increase in phosphorylation of FAK, ERK and Src kinases in U937 cells. (Subramani, J., et. al. (2013). J Immunol. 191(7):3905-12).Immunoprecipitation Analysis: A representative lot immunoprecipitated Aminopeptidase N (CD13) in Immunoprecipitation applications (Subramani, J., et. al. (2013). J Immunol. 191(7):3905-12).
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Gene Alias AP-N;hAPN;Alanyl aminopeptidase;Aminopeptidase M;AP-M;Microsomal aminopeptidase;Myeloid plasma membrane glycoprotein CD13;gp150;CD13
Gene Symbols ANPEP;APN;CD13;PEPN
Host Species Mouse
Immunogen Isolated human dermal fibroblasts.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001141
Test Specificity Clone 452 specifically detects Aminopeptidase N (CD13) in human cells.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.