Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aminopeptidase O Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552084
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552084 Supplier Invitrogen™ Supplier No. PA552084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ECSSWYYYVT MPMPASTFTI AVGCWTEMKM ETWSSNDLAT ERPFSPSEAN FRHVGVCSHM EYPCRFQNAS ATTQEIIPHR VFAPVCLTGA CQETLLRLIP PCLSAAHSVL GAHPFSRLDV LIVPANFPSL GMA Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

Aminopeptidases catalyze the hydrolysis of amino acid residues from the N-terminus of peptide or protein substrates. Able to cleave angiotensin III to generate angiotensin IV, a bioactive peptide of the renin-angiotensin pathway. Not able to cleave angiotensin I and angiotensin II. May play a role in the proteolytic processing of bioactive peptides in tissues such as testis and heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aminopeptidase O
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AOPEP
Gene Accession No. Q8N6M6
Gene Alias Aminopeptidase O; AOPEP; APO; AP-O; AP-OFLJ55832; C90RF3; C9orf3; chromosome 9 open reading frame 3; FLJ14675; FLJ40923; ONPEP
Gene Symbols AOPEP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Aminopeptidase O. Recombinant protein control fragment (Product #RP-101028).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84909
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.