Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aminopeptidase O Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567260
Description
Immunogen sequence: LFDTDTWSLQI RKTGAQTATD FPHAIRIWYK TKPEGRSVTW TSDQSGRPCV YTVGSPINNR ALFPCQEPPV AMSTWQATVR AAASFVVLMS GENSAKPT Highest antigen sequence identity to the following orthologs - mouse 86%, rat 90%.
Aminopeptidases catalyze the hydrolysis of amino acid residues from the N-terminus of peptide or protein substrates. Able to cleave angiotensin III to generate angiotensin IV, a bioactive peptide of the renin-angiotensin pathway. Not able to cleave angiotensin I and angiotensin II. May play a role in the proteolytic processing of bioactive peptides in tissues such as testis and heart.
Specifications
Aminopeptidase O | |
Polyclonal | |
Unconjugated | |
AOPEP | |
Aminopeptidase O; AOPEP; APO; AP-O; AP-OFLJ55832; C90RF3; C9orf3; chromosome 9 open reading frame 3; FLJ14675; FLJ40923; ONPEP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N6M6 | |
AOPEP | |
Recombinant Human Aminopeptidase O. Recombinant protein control fragment (Product #RP-107896). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction