Learn More
Invitrogen™ Aminopeptidase P Monoclonal Antibody (JG12), eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ BMS1104

Description
Description: Aminopeptidase P is a glycosyl-phosphatidylinositol (GPI) anchored membrane enzyme that is recognized by a monoclonal antibody designated JG12 that was raised against rat glomerular membrane protein fractions and labeled intensely microvessels of kidney and lung. Applications Tested: Immunohistochemistry (Paraffin).
This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.
Specifications
Aminopeptidase P | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O54975 | |
Xpnpep1 | |
Rec. Rat Aminopeptidase P. | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 κ |
Immunohistochemistry | |
JG12 | |
Unconjugated | |
Xpnpep1 | |
Aminoacylproline aminopeptidase; aminopeptidase P, cytosolic; App; APP1; cytoplasmic aminopeptidase P; cytosolic aminopeptidase P; D230045I08Rik; OTTHUMP00000020457; OTTHUMP00000058856; RGD:621274}; sAmp; soluble aminopeptidase P; Xaa-Pro aminopeptidase 1; XPNPEP; XPNPEP1; xpnpep1 {ECO:0000312; XPNPEPL; XPNPEPL1; X-Pro aminopeptidase 1; X-Pro aminopeptidase 1, soluble; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1; X-prolyl aminopeptidase 1, soluble | |
Mouse | |
Affinity chromatography | |
RUO | |
170751 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.