Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminopeptidase P1/XPNPEP1 Antibody (SAIC-07E-1), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320091MFV450
Description
Aminopeptidase P1/XPNPEP1 Monoclonal antibody specifically detects Aminopeptidase P1/XPNPEP1 in Human samples. It is validated for Flow CytometrySpecifications
Aminopeptidase P1/XPNPEP1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
SAIC-07E-1 | |
Flow Cytometry | |
Aminoacylproline aminopeptidase, APP1, cytosolic, Cytosolic aminopeptidase P, EC 3.4.11.9, sAmp, xaa-Pro aminopeptidase 1, XPNPEPLX-prolyl aminopeptidase (aminopeptidase P)-like, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, X-prolyl aminopeptidase 1, soluble | |
Peptide TLSLDEVYLIDSGAQYK derived from Xaa-Pro aminopeptidase 1 conjugated to KLH. | |
0.1 mL | |
metabolism, Signal Transduction | |
7511 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction