Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aminopeptidase P2/XPNPEP2 Antibody (014), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290013JF549
Description
Aminopeptidase P2/XPNPEP2 Monoclonal antibody specifically detects Aminopeptidase P2/XPNPEP2 in Human samples. It is validated for ELISASpecifications
Aminopeptidase P2/XPNPEP2 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
7512 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
014 | |
ELISA | |
Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound | |
Rabbit immunized with purified, recombinant Human Aminopeptidase P2/XPNPEP2 (Uniprot#: O43895, Met 1-Ala 650) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction