Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminopurvalanol A, Tocris Bioscience™
SDP

Catalog No. 207250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-725-0 50 mg
20-721-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-725-0 Supplier R&D Systems Supplier No. 2072/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective cdk inhibitor; potently inhibits cdk1, cdk2 and cdk5

Cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 20 - 35 nM for cdk1, cdk2 and cdk5). Also inhibits ERK1 (IC50 = 12.0 μM) and ERK2 (IC50 = 3.1 μM) and is 3000-fold selective over a range of other protein kinases (IC50 >100 μM). Arrests cell cycle at G2/M boundary (IC50 = 1.25 μM), and induces apoptosis at concentrations >10 μM. Cell permeable.

Chemical Identifiers

CAS 220792-57-4
Molecular Formula C19H26ClN7O
Molecular Weight (g/mol) 403.915
InChI Key RAMROQQYRRQPDL-UHFFFAOYSA-N
Synonym d0dm5s, aminopurvalanol a and ng-97
PubChem CID 9549304
IUPAC Name 2-[[6-(3-amino-5-chloroanilino)-9-propan-2-ylpurin-2-yl]amino]-3-methylbutan-1-ol
SMILES CC(C)C(CO)NC1=NC2=C(C(=N1)NC3=CC(=CC(=C3)N)Cl)N=CN2C(C)C

Specifications

Quantity 50 mg
Formula Weight 403.91
Percent Purity >98%
Chemical Name or Material Aminopurvalanol A
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.