Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Amintra (R) Cobalt NTA Resin
SDP

Catalog No. ACO0100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACO0100 100 mL
ACO0010 10 mL
ACO0025 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ACO0100 Supplier Novus Biologicals™ Supplier No. ACO0100

Affinity resin is designed for simple, rapid His-tagged recombinant protein purification from a cell lysate under native or denaturing conditions

  • Metal chelate affinity chromatography is a rapid one-step purification, which removes most contaminants and can achieve purities close to homogeneity.
  • The rapid purification protocols provided in this handbook along with the NTA resin for affinity chromatography permit the recovery of high levels of pure recombinant protein in minutes.
  • Large numbers of samples can be processed at the same time.
  • This product is manufactured by Expedeon Inc. and distributed by Novus Biologicals.
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Specifications

For Use With (Application) Western Blot
Quantity 100 mL
Storage Requirements Store at 4°C. Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.