Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Amintra (R) Ni-NTA Affinity Resin
SDP

Catalog No. ANN0010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ANN0010 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ANN0010 Supplier Novus Biologicals™ Supplier No. ANN0010

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • Ni-NTA affinity resin is designed for simple, rapid His-tagged recombinant protein purification from a cell lysate under native or denaturing conditions. Metal chelate affinity chromatography is a rapid one-step purification, which removes most contaminants and can achieve purities close to homogeneity.
  • Offers excellent flow properties and high protein binding capacity: in excess of 50 mg protein/mL
  • NTA is a tetradentate chelator which occupies four of the six binding sites in the coordination sphere of the nickel ion. The other two coordination sites are usually occupied by water molecules and can interact with histidine residues of the recombinant protein. This binding minimizes metal leaching during purification.
  • This product is manufactured by Expedeon Inc. and distributed by Novus Biologicals.
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Specifications

Quantity 10 mL
Storage Requirements Store at 4°C. Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.