Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Amisyn Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551960
Description
Immunogen sequence: NKKPTQASIT KVKQFEGSTS FVRRSQWMLE QLRQVNGIDP NGDSAEFDLL FENAFDQWVA STASEKCTFF QILHHTCQRY LTDRKPEFIN CQSKIMGGNS ILHSAADSVT SAVQKASQAL NERGERLGRA EEKTEDLK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
STXBP6 binds components of the SNARE complex.
Specifications
Amisyn | |
Polyclonal | |
Unconjugated | |
Stxbp6 | |
Amisyn; BC024598; C85317; FLJ39638; HSPC156; OTTHUMP00000178754; stxbp6; syntaxin binding protein 6; syntaxin binding protein 6 (amisyn); syntaxin-binding protein 6; Unknown (protein for MGC:134570); wu:fd20e06; zgc:86678 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29091 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFX7 | |
Stxbp6 | |
Recombinant protein corresponding to Human Amisyn. Recombinant protein control fragment (Product #RP-89139). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction