Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Amisyn Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518157
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518157 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518157 Supplier Invitrogen™ Supplier No. PA518157
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

STXBP6 binds components of the SNARE complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amisyn
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Stxbp6
Gene Accession No. Q8NFX7
Gene Alias Amisyn; BC024598; C85317; FLJ39638; HSPC156; OTTHUMP00000178754; stxbp6; syntaxin binding protein 6; syntaxin binding protein 6 (amisyn); syntaxin-binding protein 6; Unknown (protein for MGC:134570); wu:fd20e06; zgc:86678
Gene Symbols Stxbp6
Host Species Goat
Immunogen Synthetic peptide sequence (SAKSAISKEIFAP) corresponding to the N-terminus amino acids of STXBP6 (aa 2-15).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29091
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.