Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Amlodipine besylate
GREENER_CHOICESDP

Catalog No. 257150
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-715-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-715-0 Supplier Tocris Bioscience™ Supplier No. 2571/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CaV1.x blocker

L-type calcium channel blocker that displays antihypertensive properties. Inhibits Ca2+-induced contractions in depolarized rat aorta (IC50 = 1.9 nM) and displays vasoprotective effects in cardiovascular disease. Inhibits proliferation of human vascular smooth muscle cells and epidermoid carcinoma A431 cells (IC50 = 25 μM).

Chemical Identifiers

CAS 111470-99-6
Molecular Formula C26H31ClN2O8S

Specifications

Quantity 50 mg
Formula Weight 567.05
Purity 98%
Molecular Weight (g/mol) 567.05
Chemical Name or Material Amlodipine besylate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.