Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AMMECR1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566067
Description
Immunogen sequence: MGKRRCVPPLE PKLAAGCCGV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
AMMECR1L is a protein coding gene.
Specifications
AMMECR1L | |
Polyclonal | |
Unconjugated | |
AMMECR1L | |
AMME chromosomal region gene 1-like; AMMECR1 like; AMMECR1L; AMMECR1-like; AMMECR1-like protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
83607 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6DCA0 | |
AMMECR1L | |
Recombinant Human AMMECR1L. Recombinant protein control fragment (Product #RP-105062). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction