Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia, 0.5M solution in THF, AcroSeal™

Catalog No. ac388441000
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 7664-41-7 | H3N | 17.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonia
Color Colorless
CAS 7664-41-7, 109-99-9
Molecular Formula H3N
Linear Formula NH3
MDL Number MFCD00011418
Concentration 0.40 to 0.60M
Density 0.8500g/mL
Merck Index 15, 488
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most organic solvents
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILES N
IUPAC Name azane
Molecular Weight (g/mol) 17.03
PubChem CID 222
ChEBI CHEBI:16134
Formula Weight 17.03
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.85
Quantity 100 mL
Flash Point −36°C
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.