Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ammonia, 7M in methanol

Manufacturer:  Alfa Aesar™ H30382AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH30382AE


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7664-41-7
NH3
MFCD00011418
14,492
H3N
17.03
UN3286
Ammonia

Specifications

7664-41-7
7M in methanol
17.03
UN3286
100mL
Ammonia
H3N
NH3
MFCD00011418
14,492
14°C (57°F)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H311-H331-H370-H314
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.

RTECSNumber : BO0875000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit