Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonia-Cyanide TS, (U.S.P. Test Solution), Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 mL
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Ammonia-Cyanide TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Specifications
Specifications
CAS Max % | 3 to 5%, 1 to 3%, 80 to 100% |
UN Number | UN2922 |
Grade | Chemical Solution |
Packaging | Amber Glass Bottle |
pH | Alkaline |
Quantity | 100 mL |
Chemical Name or Material | Ammonia-Cyanide TS |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction