Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia, Household, Innovating Science™
SCI_ED

Catalog No. S000507 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000507 1 L
S000508 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning: Causes skin irritation, causes serious eye irritation, very toxic to aquatic life.
Catalog No. S000507 Supplier Aldon Corporation Supplier No. IS10071
Add to Cart
Add to Cart
Warning: Causes skin irritation, causes serious eye irritation, very toxic to aquatic life.

Household Ammonia

Laboratory Use Only

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5

Specifications

Quantity 1 L
Grade Laboratory
Packaging HDPE
Chemical Name or Material Ammonium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.