Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonia Nitrogen Standard, 500 ppm N (608 ppm NH3), Reagents
Click to view available options
Quantity:
250 mL
500 mL
Specifications
Specifications
| Chemical Name or Material | Ammonia Nitrogen Standard, 500 ppm N (608 ppm NH_3) |
| Packaging | Poly Bottle |
| CAS | 7732-18-5, 12125-02-9 |
| Specific Gravity | 1.0006 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| For Use With (Equipment) | Nitrogen Analyzer |
| Solubility | Completely miscible |
| Industry Type | Nitrogen Analysis |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction