Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonia Solution
SCI_ED

Catalog No. p-4416678 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
1 gal.
Packaging:
Solution

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7664-41-7, 7732-18-5
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
PubChem CID 222
ChEBI CHEBI:16134
IUPAC Name ammonia
SMILES N

Specifications

Color Colorless
Assay Percent Range < 5%
Density 0.98
Quantity 500 mL
Formula Weight 17.03
Packaging Solution
Physical Form Liquid
Chemical Name or Material Ammonia Solution
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.