Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonia Solution, Strong, N.F. - F.C.C., J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Specifications
Specifications
Quantity | 2.5 L |
IUPAC Name | azane |
Formula Weight | 17.03 |
Grade | FCC/NF |
Packaging | Glass Bottle |
Specific Gravity | 1L = 0.90kg |
Chemical Name or Material | Ammonia Solution, Strong |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction