Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia Standard, 100 ppm NH3 (82.2 ppm N), Ricca Chemical
SDP

Catalog No. 6131 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
613-1 4 L Natural Poly Bottle
R6134 120 mL Natural Poly Bottle
613-16 500 mL Natural Poly Bottle
613-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 613-1 Supplier Ricca Chemical Company Supplier No. 6131
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.03
CAS Max % 100.0, 0.03
Equivalence 1 mL = 0.1 mg NH3
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 4 L
Chemical Name or Material Ammonia Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.