Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia Standard, 1000 ppm NH3 (822 ppm N), Reagents

Catalog No. 01336032
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L

Catalog No. 01-336-032

Supplier: Reagents Holdings Llc CA0150004A

Only null left
Add to Cart
Add to Cart

Ammonia Standard, 1000 ppm NH3 (822 ppm N), Reagent, use for Nitrogen analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ammonia Standard, 1000 ppm NH_3 (822 ppm N)
Packaging Poly Bottle
CAS 7732-18-5, 12125-02-9
Specific Gravity 1.001
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Nitrogen Analyzer
Solubility Completely miscible
Type Conductivity Standard
Industry Type Nitrogen Analysis
Quantity 4 L
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.