Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia Standard, 55 mg/L N as NH3 (55 ppm N), Reagents

Catalog No. 01336141
Change view
Click to view available options
Quantity:
4 L
Catalog No. Quantity
01-336-141 4 L
1 options

Catalog No. 01-336-141

Supplier: Reagents Holdings Llc CA0038004C

Only null left
Add to Cart
Add to Cart

Ammonia Standard, 55 mg/L N as NH3 (55 ppm N), Reagent, use for Nitrogen analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ammonia Standard, 55 mg/L N as NH_3 (55 ppm N)
Packaging Glass Bottle
CAS 7732-18-5, 12125-02-9
Specific Gravity 1.0001
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Nitrogen Analyzer
Solubility Completely miscible
Type Conductivity Standard
Industry Type Nitrogen Analysis
Quantity 4 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.