Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammoniated Mercury, Powder, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18600555 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail

AM168, 10124-48-8, Hg(NH2)Cl

Spectrum™ Chemical Ammoniated Mercury, Powder, USP is used to treat a variety of minor skin infections and other skin disorders. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10124-48-8
Molecular Formula ClH2HgN
Molecular Weight (g/mol) 252.06
InChI Key WRWRKDRWMURIBI-UHFFFAOYSA-M
IUPAC Name mercury(2+) azanide chloride
SMILES [NH2-].[Cl-].[Hg++]

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 2.5 kg
UN Number UN1630
Percent Purity 98 to 100.5%
Grade USP
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.