Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, ACS Reagent, Reagents

Catalog No. 01338109
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
12 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-109 500 g
01-338-114 100 g
01-338-112 12 kg
01-338-111 25 kg
01-338-110 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-338-109 Supplier Reagents Holdings Llc Supplier No. C1129500500C2
Add to Cart
Add to Cart

Ammonium Acetate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.08
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym Acetic Acid ammonium salt, Ammonium ethanoate
IUPAC Name ammonium acetate
SMILES [NH4+].CC([O-])=O

Specifications

Melting Point 114°C
Color Colorless/White
pH 6.7 to 7.3
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.17
Chemical Name or Material Ammonium Acetate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.