Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate Buffer, For Iron, APHA 3500-Fe B, Certified, LabChem™

Catalog No. p-5393419
Click to view available options
Quantity:
500 mL
1 L
4 L

Specifications

Chemical Name or Material Ammonium Acetate Buffer
Name Note For Iron
CAS 64-19-7, 631-61-8, 7732-18-5
CAS Max % 68.75, 22.36, 8.89
Color Colorless
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors; Gaseous Ammonia
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Molecular Formula CH3COONH4
Linear Formula CH3COONH4
Packaging Poly Bottle
UN Number UN2790
Solubility Information Soluble in water
Formula Weight 77.08
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.