Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥98%, Solstice

Catalog No. 6002407 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
10 kg
20 kg
6 x 1 kg
6 x 2.5 kg
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-024-07 1 kg
60-024-05 100 g
60-024-10 500 g
60-024-08 2.5 kg
60-046-128 6 x 500 g
60-046-126 6 x 1 kg
60-024-06 10 kg
60-046-127 6 x 2.5 kg
60-024-09 20 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 60-024-07 Supplier Solstice Supplier No. 323011KG
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Quantity 1 kg
Linear Formula CH3CO2NH4
Beilstein 4186741
Formula Weight 77.08g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.01% (as SO4)
Chemical Name or Material Ammonium Acetate, Reag. Ph. Eur.
EINECSNumber 211-162-9
RTECSNumber AF3675000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.