Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bicarbonate, Crystal, 99%, Spectrum™ Chemical
SDP

Catalog No. 18608578 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-578 12 kg Poly Bottle
18-608-577 125 g Poly Bottle
18-608-579 500 g Poly Bottle
18-608-580 2.5 kg Poly Bottle
18-608-581 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-578 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A112512KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1066-33-7

Specifications

CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 12 kg
Molecular Weight (g/mol) 79.06
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Ammonium Bicarbonate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.