Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bicarbonate (Powder/Certified), Fisher Chemical™

Catalog No. A64350 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fiber Drum
Poly Bottle

Chemical Identifiers

CAS 1066-33-7
Molecular Formula CH5NO3
Molecular Weight (g/mol) 79.06
MDL Number MFCD00012138
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
IUPAC Name carbonic acid amine
SMILES N.OC(O)=O

Specifications

Melting Point 60°C
Color White
pH 7.8
Physical Form Solid
Quantity 50 kg
Assay Percent Range 21.30 to 21.73 %
Linear Formula NH4HCO3
Formula Weight 79.06
Percent Purity 21.30 to 21.73%
Grade Certified
Identification Pass Test
Packaging Fiber Drum
Vapor Pressure 60mmHg at 25°C
Density 1.586g/cm³
Chemical Name or Material Ammonium Bicarbonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.