Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Bifluoride (Crystalline/Technical), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
Specifications
Specifications
Boiling Point | 230°C |
Melting Point | 125°C |
Color | White |
pH | 3.5 |
Physical Form | Solid |
Quantity | 3 kg |
UN Number | UN 1727 |
Formula Weight | 57.04 |
Grade | Technical |
Identification | Pass Test |
Show More |
Safety and Handling

DOTInformation | DOT Class 8, : Corrosive |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction