Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium bromide, 99+%, for analysis

Catalog No. p-4509821
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle

CAS: 12124-97-9 | BrH4N | 97.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12124-97-9
Molecular Formula BrH4N
Molecular Weight (g/mol) 97.94
MDL Number MFCD00011419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
IUPAC Name ammonium bromide
SMILES [NH4+].[Br-]

Specifications

Boiling Point 235.0°C
Melting Point 452.0°C
Color White
pH 4.5 to 6.0 (5% soln.)
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99% min. (on dry substance) (Argentometry)
Linear Formula NH4Br
Merck Index 15,502
Solubility Information Solubility in water: 970g/L (25°C). Other solubilities: 1456g/L (100°C), freely soluble in alcohol and acetone, slightly soluble in ether, practically insoluble in ethyl acetate
Formula Weight 97.94
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.2% (105°C) max.
Packaging Glass bottle
Chemical Name or Material Ammonium bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 235-183-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.